Order Entry
Switzerland
ContactUsLinkComponent
 
Organic reference standard, 2,3,4,5,6-Pentachlorobiphenyl (PCB No. 116) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-116
 :  
Organic reference standard, 2,3,4,5,6-Pentachlorobiphenyl (PCB No. 116) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-116
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    18259-05-7
  • Description:
    2,3,4,5,6-Pentachlorobiphenyl (PCB No. 116) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,3,4,5,6-Pentachlorobiphenyl (PCB No. 116)