Organic reference standard, 2,3,3',4,4',5,5',6-Octachlorobiphenyl (PCB No. 205) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20020500IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:74472-53-0
- Description:2,3,3',4,4',5,5',6-Octachlorobiphenyl (PCB No. 205) 10 µg/ml in isooctane
- Formula:C₁₂H₂Cl₈
- Description:2,3,3',4,4',5,5',6-Octachlorobiphenyl (PCB No. 205)