Order Entry
Switzerland
ContactUsLinkComponent
L(+)-Tartaric acid, granular NF, Multi-Compendial, J.T.Baker®
L(+)-Tartaric acid, granular NF, Multi-Compendial, J.T.Baker®
Catalog # 4105-05
Supplier:  Avantor
CAS Number:  
undefined
L(+)-Tartaric acid, granular NF, Multi-Compendial, J.T.Baker®
Catalog # 4105-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Specific Rotation [Å]²⁵^D(calculated on dried basis)(+):
    12.0 - 13.0 Degree
  • NF - Loss on Drying:
    ≤0.5 %
  • NF - Residue on Ignition:
    ≤0.1 %
  • NF - Oxalate (C₂O₄):
    Passes Test
  • NF - Sulfate (SO₄):
    Passes Test
  • NF - Assay (HOCO(CHOH)₂COOH) (dried basis):
    99.7 - 100.5 %
  • NF - Endotoxin Concentration (EU/g):
    ≤10
  • EP/BP - Assay (HOCO(CHOH)₂COOH) (dried basis):
    99.5 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Specific Rotation [Å]²º^D (+):
    12.0 - 12.8 Degree
  • EP/BP - Oxalic Acid:
    ≤360 ppm
  • EP/BP - Chloride (Cl):
    ≤100 ppm
  • EP/BP - Sulfate (SO₄):
    ≤150 ppm
  • EP/BP - Calcium (Ca):
    ≤200 ppm
  • EP/BP - Loss on Drying at 105°C:
    ≤0.2 %
  • EP/BP - Ash (sulfated):
    ≤0.1 %
  • JP - Assay (HOCO(CHOH)₂COOH) (dried basis):
    99.7 - 101.0 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Sulfate (SO₄):
    ≤0.048 %
  • JP - Oxalate (C₂O₄):
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤10 ppm
  • JP - Calcium (Ca):
    Passes Test
  • JP - Arsenic (As):
    ≤1 ppm
  • JP - Loss on Drying:
    ≤0.5 %
  • JP - Residue on Ignition:
    ≤0.05 %
  • No Class 1,2,3 or other solvents are used or produced in the:
    = 0.05 %

Specification Test Results

About this item

 :
Merck Index:13,09158
Boiling Pt:399 °C (1013 hPa)
Melting Pt:168…170 °C
Density:1.759 g/cm³ (20 °C)
Flash Pt:150 °C (closed cup)
 :150.09 g/mol
Storage Temperature:Ambient
EINECS:201-766-0
REACH:01-2119537204-47
 :GHS05
CAS Number:87-69-4
Formula:C₄H₆O₆
MDL Number:MFCD00064207