Organic reference standard, 2,3,6-Trichlorobiphenyl (PCB No. 24) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-24
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:55702-45-9
- Description:2,3,6-Trichlorobiphenyl (PCB No. 24) 35 µg/ml in isooctane
- Environmentally Preferable:
- Formula:C₁₂H₇Cl₃
- Description:2,3,6-Trichlorobiphenyl (PCB No. 24)