Organic reference standard, 2,3,4,4',5-Pentachlorobiphenyl (PCB No. 114) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20011400IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:74472-37-0
- Description:2,3,4,4',5-Pentachlorobiphenyl (PCB No. 114) 10 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3,4,4',5-Pentachlorobiphenyl (PCB No. 114)