Organic reference standard, 2,3',4-Trichlorobiphenyl (PCB No. 25) 35 µg/ml in isooctane, CIL
Katalognr. PROOCIL-PCB-25
Anbieter: LGC Standards PROMOCHEM
CAS-Nummer:
Technische Daten
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:55712-37-3
- Description:2,3',4-Trichlorobiphenyl (PCB No. 25) 35 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,3',4-Trichlorobiphenyl (PCB No. 25)