Order Entry
Switzerland
ContactUsLinkComponent
 
Organic reference standard, 2,3,3',5,5'-Pentachlorobiphenyl (PCB No. 111) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-111
 :  
Organic reference standard, 2,3,3',5,5'-Pentachlorobiphenyl (PCB No. 111) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-111
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    39635-32-0
  • Description:
    2,3,3',5,5'-Pentachlorobiphenyl (PCB No. 111) 35 µg/ml in isooctane
  • Environmentally Preferable:
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,3,3',5,5'-Pentachlorobiphenyl (PCB No. 111)