Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

D-(-)-Glutamic acid

Marketsource
Supplier:   BACHEM BIOCHEMICA
D-(-)-Glutamic acid
Supplier:  BACHEM BIOCHEMICA
Marketsource
Certificates
D-(-)-Glutamic acid

2 Options Available Below

Product Details & Documents
Synonyms:  (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
  • CAS Number:6893-26-1
  • Formula:C₅H₉NO₄
  • MDL Number:MFCD00063112
  • Molecular Weight:147.13 g/mol
  • Merck Index:13,04482
  • Boiling Pt:325 °C (1013 hPa)
  • Melting Pt:213 °C
  • Density:1.533 g/cm³ (20 °C)
  • EINECS:230-000-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.