Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 2,3,3',4,5,5'-Hexachlorobiphenyl (PCB No. 159) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,3,3',4,5,5'-Hexachlorobiphenyl (PCB No. 159) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-159
 :  
Organic reference standard, 2,3,3',4,5,5'-Hexachlorobiphenyl (PCB No. 159) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-159
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    39635-35-3
  • Description:
    2,3,3',4,5,5'-Hexachlorobiphenyl (PCB No. 159) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,3,3',4,5,5'-Hexachlorobiphenyl (PCB No. 159)