D-(-)-Glutamic acid 99+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
- CAS Number:6893-26-1
- Formula:C₅H₉NO₄
- MDL Number:MFCD00063112
- Molecular Weight:147.13 g/mol
- Merck Index:13,04482
- Boiling Pt:325 °C (1013 hPa)
- Melting Pt:200…202 °C
- Density:1,533 g/cm³ (20 °C)
- EINECS:230-000-8
3 Options Available Below
Product Details & Documents
Synonyms: (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
- CAS Number:6893-26-1
- Formula:C₅H₉NO₄
- MDL Number:MFCD00063112
- Molecular Weight:147.13 g/mol
- Merck Index:13,04482
- Boiling Pt:325 °C (1013 hPa)
- Melting Pt:200…202 °C
- Density:1,533 g/cm³ (20 °C)
- EINECS:230-000-8
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 3 hrs)
- Sulfated ash:≤0.1 %
- HPLC:≤1 % L-content
- Specific optical rotation:-30.5° to -32.2° (20°C, 589 nm) (c=10, 2 N HCl)
- Titration with NaOH:99.0 to 101.0 %
- Other amino acids:≤0.5 % (TLC)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



