Organic reference standard, 2,3',4,4',6-Pentachlorobiphenyl (PCB No. 119) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20011900IO
Anbieter: EHRENSTORFER
CAS-Nummer:
Technische Daten
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:56558-17-9
- Description:2,3',4,4',6-Pentachlorobiphenyl (PCB No. 119) 10 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3',4,4',6-Pentachlorobiphenyl (PCB No. 119)