Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-156
CAS Number:  
Organic reference standard, 2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-156
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    38380-08-4
  • Description:
    2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156)

Specifications

About this item