Organic reference standard, 2,3',4'-Trichlorobiphenyl (PCB No. 33) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20003300IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:38444-86-9
- Description:2,3',4'-Trichlorobiphenyl (PCB No. 33) 10 µg/ml in isooctane
- Formula:C₁₂H₇Cl₃
- Description:2,3',4'-Trichlorobiphenyl (PCB No. 33)