Order Entry
Switzerland
ContactUsLinkComponent
 
Organic reference standard, 2,2',3,4,5-Pentachlorobiphenyl (PCB No. 86) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-86
 :  
Organic reference standard, 2,2',3,4,5-Pentachlorobiphenyl (PCB No. 86) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-86
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    55312-69-1
  • Description:
    2,2',3,4,5-Pentachlorobiphenyl (PCB No. 86) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,2',3,4,5-Pentachlorobiphenyl (PCB No. 86)