Organic reference standard, 2,2',3,6,6'-Pentachlorobiphenyl (PCB No. 96) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-96
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:73575-54-9
- Description:2,2',3,6,6'-Pentachlorobiphenyl (PCB No. 96) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,2',3,6,6'-Pentachlorobiphenyl (PCB No. 96)