Organic reference standard, 2,3',4,4',5'-Pentachlorobiphenyl (PCB No. 123) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20012300IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:65510-44-3
- Description:2,3',4,4',5'-Pentachlorobiphenyl (PCB No. 123) 10 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3',4,4',5'-Pentachlorobiphenyl (PCB No. 123)