Order Entry
Switzerland
ContactUsLinkComponent
 
Organic reference standard, 3,5-Dichlorobiphenyl (PCB No. 14) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20001400IO
 :  
Organic reference standard, 3,5-Dichlorobiphenyl (PCB No. 14) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20001400IO
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    34883-41-5
  • Description:
    3,5-Dichlorobiphenyl (PCB No. 14) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₈Cl₂
  • Description:
    3,5-Dichlorobiphenyl (PCB No. 14