Organischer Referenzstandard, 2,3',4,4'-Tetrachlorbiphenyl (PCB Nr. 66) 10 µg/ml in Isooctan
Catalog # EHERL20006600IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Packung:Glasflasche
- VE:10 mL
- CAS Nr.:32598-10-0
- Bezeichnung:2,3',4,4'-Tetrachlorbiphenyl (PCB Nr. 66) 10 µg/ml in Isooctan
- Formel:C₁₂H₆Cl₄
- Bezeichnung:2,3',4,4'-Tetrachlorobiphenyl (PCB No. 66)