Order Entry
Switzerland
ContactUsLinkComponent
 
Organic reference standard, 2,2',4,4'-Tetrachlorobiphenyl (PCB No. 47) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-47
CAS Number:  
Organic reference standard, 2,2',4,4'-Tetrachlorobiphenyl (PCB No. 47) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-47
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    2437-79-8
  • Description:
    2,2',4,4'-Tetrachlorobiphenyl (PCB No. 47) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    2,2',4,4'-Tetrachlorobiphenyl (PCB No. 47)

Specifications

About this item