Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 2,2',3,3',6,6'-Hexachlorobiphenyl (PCB No. 136) 10 µg/ml in isooctane, Dr Ehrenstorfer
Organic reference standard, 2,2',3,3',6,6'-Hexachlorobiphenyl (PCB No. 136) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20013600IO
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2,2',3,3',6,6'-Hexachlorobiphenyl (PCB No. 136) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20013600IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    38411-22-2
  • Description:
    2,2',3,3',6,6'-Hexachlorobiphenyl (PCB No. 136) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,2',3,3',6,6'-Hexachlorobiphenyl (PCB No. 136)

Specifications

About this item