Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 2,2',3,3',5,5'-Hexachlorobiphenyl (PCB No. 133) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,3',5,5'-Hexachlorobiphenyl (PCB No. 133) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-133
CAS Number:  
Organic reference standard, 2,2',3,3',5,5'-Hexachlorobiphenyl (PCB No. 133) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-133
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    35694-04-3
  • Description:
    2,2',3,3',5,5'-Hexachlorobiphenyl (PCB No. 133) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    2,2',3,3',5,5'-Hexachlorobiphenyl (PCB No. 133)

Specifications

About this item