Organic reference standard, 2,2',3,4,4',6'-Hexachlorobiphenyl (PCB No. 140) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-140
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:59291-64-4
- Description:2,2',3,4,4',6'-Hexachlorobiphenyl (PCB No. 140) 35 µg/ml in isooctane
- Environmentally Preferable:
- Formula:C₁₂H₄Cl₆
- Description:2,2',3,4,4',6'-Hexachlorobiphenyl (PCB No. 140)