Order Entry
Switzerland
ContactUsLinkComponent
 
Organic reference standard, 2,2',3,3',4,6,6'-Heptachlorobiphenyl (PCB No. 176) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-176
 :  
Organic reference standard, 2,2',3,3',4,6,6'-Heptachlorobiphenyl (PCB No. 176) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-176
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    52663-65-7
  • Description:
    2,2',3,3',4,6,6'-Heptachlorobiphenyl (PCB No. 176) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₃Cl₇
  • Description:
    2,2',3,3',4,6,6'-Heptachlorobiphenyl (PCB No. 176)