Organic reference standard, 2,3,3',4,4',5,5'-Heptachlorobiphenyl (PCB No. 189) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20018900IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:39635-31-9
- Description:2,3,3',4,4',5,5'-Heptachlorobiphenyl (PCB No. 189) 10 µg/ml in isooctane
- Formula:C₁₂H₃Cl₇
- Description:2,3,3',4,4',5,5'-Heptachlorobiphenyl (PCB No. 189)