Organic reference standard, 2,2',3,4,4',5,6'-Heptachlorobiphenyl (PCB No. 182) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-182
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:60145-23-5
- Description:2,2',3,4,4',5,6'-Heptachlorobiphenyl (PCB No. 182) 35 µg/ml in isooctane
- Environmentally Preferable:
- Formula:C₁₂H₃Cl₇
- Description:2,2',3,4,4',5,6'-Heptachlorobiphenyl (PCB No. 182)