Pesticide reference standard, 2,3,6-Trichloroanisole 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17713600CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:50375-10-5
- Description:2,3,6-Trichloroanisole 10 µg/ml in cyclohexane
- Formula:C₇H₅Cl₃O
- Description:2,3,6-Trichloroanisole