Organic reference standard, Tetrachloroethylene 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA17358300ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:127-18-4
- Description:Tetrachloroethylene 100 µg/ml in methanol
- Formula:C₂Cl₄
- Description:Tetrachloroethene