Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Gentamicin sulphate solution 10 mg/ml

 :   Biowest
Gentamicin sulphate solution 10 mg/ml
 :  Biowest
 
Gentamicin sulphate solution 10 mg/ml
Gentamicin sulphate solution 10 mg/ml

 

 

Gentamicin sulfate acts against gram-negative, gram-positive bacteria and mycoplasma.

  • Storage temperature of –20 °C
  • pH of 4,5±0,3
  • Shelf life of 24 months
  • Endotoxin: <10 EU/ml (<1 ng/ml)

Used in cell culture applications at 5 ml/l. This concentration is for tissue culture media containing serum, serum-free media generally require lower concentration.

 : This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

 :  NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  •  :1405-41-0
  •  :MFCD00270181

 

Product Family Options

 

  • Pk
     
     
  • 10 mL
       
       
    1 * 10 mL
    1 * 10 mL - SFr. 16.10
    SFr. 16.10
    1 * 10 mL
       SFr. 16.10

  • 100 ml
       
       
    1 * 100 mL
    1 * 100 mL - SFr. 44.10
    SFr. 44.10
    1 * 100 mL
       SFr. 44.10