Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
Nickel(II) sulphate hexahydrate, EMSURE® for analysis, Supelco®
 :  Merck
 
 
 
 
Nickel(II) sulphate hexahydrate, EMSURE® for analysis, Supelco®
 :  Merck
 

 

  • Solubility:
    650 g/l (20 °C)
  • Melting Point:
    53 °C
  • Molar Mass:
    262.86 g/mol
  • Bulk Density:
    1000 kg/m3
  • Density:
    2.07 g/cm3 (20 °C)
  • pH:
    4.3 - 4.7 (100 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

 

Frequently Bought Together