


Nickel(II) sulphate hexahydrate ≥99.97% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10101-97-0
:
Formula:NiSO₄·6H₂O
MDL Number:MFCD00149813
:262.85 g/mol
Merck Index:14,6517
Melting Pt:840 °C
Density:2.070
EINECS:232-104-9
UN:3288
ADR:6.1,III
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together