Order Entry
Switzerland
ContactUsLinkComponent
Iron(II) sulphate heptahydrate ≥96%, GPR RECTAPUR®
Irritant
Iron(II) sulphate heptahydrate ≥96%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Iron(II) sulphate heptahydrate ≥96%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 96 %
  • Not precipitated by NH4OH (as SO4):
    Max. 0.5 %
  • Cl (Chloride):
    Max. 50 ppm

Specification Test Results

Frequently Bought Together