




Nickel(II) nitrate hexahydrate ≥96%, GPR RECTAPUR®
Catalog # 25871.292
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:Min. 96 %
- Cl (Chloride):Max. 0.02 %
- SO4 (Sulphate):Max. 0.05 %
- Cd (Cadmium):Max. 25 ppm
- Cu (Copper):Max. 25 ppm
- Pb (Lead):Max. 25 ppm
- Zn (Zinc):Max. 25 ppm
Specification Test Results
About this item
:
:290.79 g/mol
Merck Index:12,06596
Boiling Pt:137 °C (1013 hPa)
Melting Pt:56.7 °C
Density:2.05 g/cm³ (20 °C)
EINECS:236-068-5
UN:2725
ADR:5.1,III
:GHS03
:GHS05
:GHS08
:GHS07
:GHS09
CAS Number:13478-00-7
Formula:Ni(NO₃)₂·6H₂O
MDL Number:MFCD00149805