




Nickel(II) nitrate hexahydrate, EMSURE® for analysis, Supelco®
: Merck
:13478-00-7
:
:Ni(NO₃)₂·6H₂O
:MFCD00149805
:290.79 g/mol
:12,06596
:137 °C (1013 hPa)
:56.7 °C
:2.05 g/cm³ (20 °C)
:2725
:5.1,III
:M-Clarity™ quality level = MQ300
:236-068-5
:GHS03
:GHS05
:GHS08
:GHS07
:GHS09
- Solubility:940 g/l (20 °C)
- Melting Point:56.7 °C
- Molar Mass:290.81 g/mol
- Bulk Density:800 kg/m3
- Boiling Point:136.7 °C (1013 hPa)
- Density:2.05 g/cm3 (20 °C)
- pH:5 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Frequently Bought Together