Order Entry
Switzerland
ContactUsLinkComponent
Cobalt(II) nitrate hexahydrate ≥98%, GPR RECTAPUR®
Health
Environment
Cobalt(II) nitrate hexahydrate ≥98%, GPR RECTAPUR®
Catalog # 22912.237
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Environment
Cobalt(II) nitrate hexahydrate ≥98%, GPR RECTAPUR®
Catalog # 22912.237
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Cu (Copper):
    Max. 100 ppm
  • Fe (Iron):
    Max. 100 ppm
  • Ni (Nickel):
    Max. 0.6 %
  • Pb (Lead):
    Max. 100 ppm

Specification Test Results

About this item

 :
 :291.03 g/mol
Merck Index:13,02469
Melting Pt:57 °C
Density:1.88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-402-1
UN:1477
ADR:5.1,III
 :GHS08
 :GHS09
CAS Number:10026-22-9
Formula:Co(NO₃)₂·6H₂O
MDL Number:MFCD00149647