Order Entry
Switzerland
ContactUsLinkComponent
Cobalt(II) nitrate hexahydrate ≥96%, TECHNICAL
Health
Environment
Cobalt(II) nitrate hexahydrate ≥96%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Health
Environment
Cobalt(II) nitrate hexahydrate ≥96%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 96 %

Specification Test Results

Frequently Bought Together