

Iron(III) chloride hexahydrate 97% ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:10025-77-1
:
Formula:FeCl₃·6H₂O
MDL Number:MFCD00149712
:270.3 g/mol
Merck Index:13,04048
Boiling Pt:280…285 °C
Melting Pt:37 °C
Density:2.8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-729-4
UN:3260
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Assay:97.0 - 102.0 %
- Insoluble in acid:Max. 0.01 %
- Calcium (Ca):Max. 0.01 %
- Copper (Cu):Max. 0.003 %
- Potassium (K):Max. 0.005 %
- Magnesium (Mg):Max. 0.005 %
- Sodium (Na):Max. 0.05 %
- Zinc (Zn):Max. 0.003 %
- Iron (Fe 2⁺):Max. 0.002 %
- Nitrate (NO₃):Max. 0.01 %
- Total phosphorus (as PO₄):Max. 0.01 %
- Sulfate (SO₄):Max. 0.01 %
Specification Test Results
Frequently Bought Together