Order Entry
Switzerland
ContactUsLinkComponent
Iron(III) chloride hexahydrate, EMSURE® for analysis, Supelco®
Corrosive
Irritant
Iron(III) chloride hexahydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Corrosive
Irritant
Iron(III) chloride hexahydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (iodometric, FeCl₃·6H₂O):
    99.0 - 102.0 %
  • Identity:
    passes test
  • Insoluble matter:
    ≤ 0.01 %
  • Free chlorine:
    passes test
  • Acidic substances:
    passes test
  • Nitrate (NO₃):
    ≤ 0.01 %
  • Sulfate (SO₄):
    ≤ 0.01 %
  • Total nitrogen (N):
    ≤ 0.001 %
  • Total phosphorus (as PO₄):
    ≤ 0.01 %
  • Heavy metals (as Pb):
    ≤ 0.005 %
  • Ca (Calcium):
    ≤ 0.01 %
  • Cu (Copper):
    ≤ 0.003 %
  • Fe II (Iron II):
    ≤ 0.002 % (The Fe(II)-content increases if the recommended storage conditions (+15 to +25 °C) are not observed)
  • K (Potassium):
    ≤ 0.005 %
  • Mg (Magnesium):
    ≤ 0.005 %
  • Na (Sodium):
    ≤ 0.05 %
  • Zn (Zinc):
    ≤ 0.003 %

Specification Test Results

Related Information

Frequently Bought Together