Order Entry
Switzerland
ContactUsLinkComponent
Cobalt(II) chloride hexahydrate 98% ACS, Fluka™
Health
Irritant
Environment
Cobalt(II) chloride hexahydrate 98% ACS, Fluka™
Certificates
Health
Irritant
Environment
Cobalt(II) chloride hexahydrate 98% ACS, Fluka™
Certificates

Specification Test Results

  • Assay:
    98.0 - 102.0 %
  • Water insoluble matter:
    Max. 0.01 %
  • Calcium (Ca):
    Max. 0.005 %
  • Copper (Cu):
    Max. 0.002 %
  • Iron (Fe):
    Max. 0.005 %
  • Potassium (K):
    Max. 0.01 %
  • Magnesium (Mg):
    Max. 0.005 %
  • Sodium (Na):
    Max. 0.05 %
  • Nickel (Ni):
    Max. 0.1 %
  • Zinc (Zn):
    Max. 0.03 %
  • Nitrate (NO₃):
    Max. 0.01 %
  • Sulfate (SO₄):
    Max. 0.01 %

Specification Test Results

Related Information

Frequently Bought Together