Order Entry
Switzerland
ContactUsLinkComponent
Manganese(II) sulphate monohydrate ≥98%, GPR RECTAPUR®
Health
Environment
Manganese(II) sulphate monohydrate ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Health
Environment
Manganese(II) sulphate monohydrate ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Heavy metals (as Pb):
    Max. 50 ppm
  • Assay:
    Min. 98 %
  • Not precipitated by (NH4)2S (as SO4):
    Max. 0.5 %
  • Cl (Chloride):
    Max. 50 ppm
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

Frequently Bought Together