



Manganese(II) chloride tetrahydrate 99+% for analysis
:13446-34-9
:
:MnCl₂·4H₂O
:MFCD00149792
:197.9 g/mol
:13,05751
:1190 °C (1013 hPa)
:58 °C
:2,01 g/cm³ (20 °C)
:231-869-6
:3077
:9,III
:GHS08
:GHS05
:GHS07
:GHS09
:Ambient
- Appearance:Pink Crystalline powder
- Titration Complexometric:≥99.0 %
- Sulfate (SO4):≤50 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤5 ppm
- Lead (Pb):≤5 ppm
- Magnesium (Mg):≤50 ppm
- Zinc (Zn):≤10 ppm
- KMnO4-reducing substance:≤0.0005 % (as O)
Frequently Bought Together