


Chromium(III) chloride hexahydrate ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10060-12-5
:
Formula:CrCl₃·6H₂O
MDL Number:MFCD00149660
:266.45 g/mol
Merck Index:14,2222
Melting Pt:83 °C
Density:1.835
EINECS:233-038-3
UN:3260
ADR:8,III
:GHS08
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together