Order Entry
Switzerland
ContactUsLinkComponent
Zinc sulphate heptahydrate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Irritant
Corrosive
Environment
Zinc sulphate heptahydrate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Environment
Zinc sulphate heptahydrate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    ≥ 99.5 %
  • Solution in water:
    Passes test
  • pH (25°C; 5 %):
    4.4 - 6.0
  • Insolubility in water:
    ≤ 100 ppm
  • Cl (Chloride):
    ≤ 5 ppm
  • Total N (Nitrogen):
    ≤ 5 ppm
  • As (Arsenic):
    ≤ 0.5 ppm
  • Ca (Calcium):
    ≤ 10 ppm
  • Cd (Cadmium):
    ≤ 5 ppm
  • Cu (Copper):
    ≤ 5 ppm
  • Fe (Iron):
    ≤ 5 ppm
  • K (Potassium):
    ≤ 10 ppm
  • Li (Lithium):
    ≤ 10 ppm
  • Mn (Manganese):
    ≤ 3 ppm
  • Na (Sodium):
    ≤ 10 ppm
  • Pb (Lead):
    ≤ 20 ppm
  • Sr (Strontium):
    ≤ 5 ppm
  • Conforms to BDH 10299:
    Passes test

Specification Test Results

Frequently Bought Together