Zinc sulphate heptahydrate, granular USP, FCC, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7446-20-0
:
Formula:ZnSO₄·7H₂O
MDL Number:MFCD00149894
Merck Index:12,10293
Melting Pt:100 °C
Density:1.97 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:9,III
:Management of Change (MOC) category = R
:287.54 g/mol
EINECS:231-793-3
UN:3077
:GHS05
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Food GMP Manufactured Product:99.0 - 108.7%
- Meets U.S.P Requirements:Passes Test
- Meets F.C.C. Requirements:Passes Test
- GMP Manufactured Product:Passes Test
- CAUTION: For Manufacturing, processing or repackaging:≤0.5%
- Bulk Food Chemical:≤3 ppm
- Bulk Pharmaceutical Chemical:≤0.002%
- USP - Assay (ZnSO₄·7H₂O):99.0 - 108.7 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Acidity:Passes Test
- USP - Alkalies and Alkaline Earths:≤ 0.9 %
- USP - Arsenic (As):≤ 3 ppm
- USP - Lead (Pb) (USP):≤ 0.002 %
- FCC - Assay (ZnSO₄·7H₂O):99.0 - 108.7 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Acidity:Passes Test
- FCC - Alkalies and Alkaline Earths:≤ 0.5 %
- FCC - Cadmium (Cd):≤ 2 mg/kg
- FCC - Lead (Pb) (FCC):≤ 4 mg/kg
- FCC - Selenium (Se):≤ 0.003 %
- FCC - Mercury (Hg):≤ 5 mg/kg
- FCC - Assay (dried basis):55.6 - 61.0 %
Specification Test Results
Frequently Bought Together