Order Entry
Switzerland
ContactUsLinkComponent
Copper(II) sulphate pentahydrate ≥98.0%, Reagent Grade
Irritant
Environment
Copper(II) sulphate pentahydrate ≥98.0%, Reagent Grade
Catalog # C7631-500G
CAS Number:  
Irritant
Environment
Copper(II) sulphate pentahydrate ≥98.0%, Reagent Grade
Catalog # C7631-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 98.0 %
  • Iron (Fe):
    Max. 0.01 %

Specification Test Results

About this item

 :
 :249.69 g/mol
Merck Index:13,02682
Boiling Pt:Decomposes on heating
Boiling Pt: Dec. (on heating)
Melting Pt:110 °C
Density:2.28 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-847-6
ADR:9,III
REACH:01-2119520566-40
 :GHS07
 :GHS09
CAS Number:7758-99-8
Formula:CuSO₄·5H₂O
MDL Number:MFCD00149681