Order Entry
Switzerland
ContactUsLinkComponent
Copper(II) sulphate pentahydrate 99.0-102.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Irritant
Corrosive
Environment
Copper(II) sulphate pentahydrate 99.0-102.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Environment
Copper(II) sulphate pentahydrate 99.0-102.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    99.0 - 102.0 %
  • Insolubility in water:
    Max. 50 ppm
  • Total N (Nitrogen):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • Ca (Calcium):
    Max. 50 ppm
  • Fe (Iron):
    Max. 20 ppm
  • K (Potassium):
    Max. 10 ppm
  • Mg (Magnesium):
    Max. 50 ppm
  • Na (Sodium):
    Max. 50 ppm
  • Ni (Nickel):
    Max. 50 ppm
  • Pb (Lead):
    Max. 50 ppm
  • Zn (Zinc):
    Max. 0.03 %
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together