

Copper(II) sulphate pentahydrate 98.0-102.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-99-8
:
Formula:CuSO₄·5H₂O
MDL Number:MFCD00149681
:249.69 g/mol
Merck Index:14,2653
Density:2.270
Storage Temperature:Ambient
EINECS:231-847-6
UN:3288
ADR:6.1,III
REACH:01-2119520566-40
:GHS06
:GHS07
Related Information
Frequently Bought Together