Order Entry
Switzerland
ContactUsLinkComponent
 
 
Ammonium iron(III) sulphate dodecahydrate 99.0-101.0%, AnalaR NORMAPUR® analytical reagent
 
undefined
 
Ammonium iron(III) sulphate dodecahydrate 99.0-101.0%, AnalaR NORMAPUR® analytical reagent
 

 

  • Assay:
    99.0 - 101.0 %
  • Insolubility in water:
    Max. 50 ppm
  • Substances not precipitated by NH4OH:
    Max. 0.1 %
  • Cl (Chloride):
    Max. 10 ppm
  • NO3 (Nitrate):
    Max. 100 ppm
  • Cu (Copper):
    Max. 20 ppm
  • Fe (II) (Iron):
    Max. 10 ppm
  • Mn (Manganese):
    Max. 50 ppm
  • Pb (Lead):
    Max. 20 ppm
  • Zn (Zinc):
    Max. 30 ppm

 

Frequently Bought Together