Magnesium sulphate heptahydrate 98.0-102.0%, powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:10034-99-8
:
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785
:246.48 g/mol
Merck Index:13,05715
Melting Pt:1124 °C
Density:2.66 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
Specification Test Results
- Assay:98,0 - 102,0%
- pH:5,0 - 8,2 (5% aq soln)
- Insoluble Matter:≤0,005%
- Chloride (Cl) :≤5 ppm
- Nitrate (NO₃):≤0,002%
- Ammonium (NH₄):≤0,002%
- Calcium (Ca):≤0,02%
- Manganese (Mn) :≤5 ppm
- Potassium (K):≤0,005%
- Sodium (Na):≤0,005%
- Strontium (Sr):≤0,005%
- Heavy Metals (Pb):≤5 ppm
- Iron (Fe):≤5 ppm
Specification Test Results
Frequently Bought Together