Calcium sulphate dihydrate 98+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10101-41-4
:
Formula:CaSO₄·2H₂O
MDL Number:MFCD00149625
:172.17 g/mol
Merck Index:13,01711
Density:2,32 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-900-3
Specification Test Results
- Appearance:White Powder
- Titration Complexometric:98.0 to 102.0 %
- Chloride (Cl):≤50 ppm
- Nitrate (NO3):≤50 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤20 ppm
- Insoluble matter:≤0.05 % (in dilute HCl)
Specification Test Results
Frequently Bought Together