Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Capping Reagent
Capping Reagent B2 60% (v/v) Lutidine in Acetonitrile, Novabiochem®
Capping Reagent B2 60% (v/v) Lutidine in Acetonitrile, Novabiochem®
Capping Reagent B2 60% (v/v) Lutidine in Acetonitrile, Novabiochem®
MERC8.57006.0500
:
Merck
:
Capping Reagent B2 60% (v/v) Lutidine in Acetonitrile, Novabiochem®
MERC8.57006.0500
:
Merck
:
Pk:
500 mL
Seq:
2725
:
:
0.87 g/cm³ (20 °C)
:
10.0 °C
:
3,II
:
M-Clarity™ quality level = MQ200
:
1993
:
GHS02
:
GHS07